3-Bromo-N-cyclopentyl-5-nitrobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4376-5G CA-4376-1G
COBBCA-4376-5GEA 0 GBP
COBBCA-4376-5G COBBCA-4376-1G
3-Bromo-N-cyclopentyl-5-nitrobenzamide
3-Bromo-N-cyclopentyl-5-nitrobenzamide
Formula: C₁₂H₁₃BrN₂O₃
MDL Number: MFCD09475860
CAS Number: 941294-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR