2-Amino-2-(2-bromophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-DL-2-Phg(2-Br)-Oh

AS24148G1 AS24148G5
ACTIAS24148G1EA 0 GBP
ACTIAS24148G1 ACTIAS24148G5
2-Amino-2-(2-bromophenyl)acetic acid
2-Amino-2-(2-bromophenyl)acetic acid
Formula: C₈H₈BrNO₂
MDL Number: MFCD02662413
CAS Number: 254762-66-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR