2-Bromo-5-nitrobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-5-nitrobenzenecarbaldehyde

AS34466G1 AS34466G5
ACTIAS34466G1EA 0 GBP
ACTIAS34466G1 ACTIAS34466G5
2-Bromo-5-nitrobenzaldehyde
2-Bromo-5-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
MW: 230.0200 g/mol
MDL Number: MFCD00462865
CAS Number: 84459-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR