5-Benzylcarbamoyl-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-(Benzylcarbamoyl)-2-fluorophenyl)boronic acid

BB-3347-1G BB-3347-5G
COBBBB-3347-1GEA 0 GBP
COBBBB-3347-1G COBBBB-3347-5G
5-Benzylcarbamoyl-2-fluorophenylboronic acid
5-Benzylcarbamoyl-2-fluorophenylboronic acid
Formula: C₁₄H₁₃BFNO₃
MW: 273.0700 g/mol
MDL Number: MFCD08436031
CAS Number: 874289-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR