2-Chloro-(4-ethoxycarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3531-5G BB-3531-1G
COBBBB-3531-5GEA 0 GBP
COBBBB-3531-5G COBBBB-3531-1G
2-Chloro-(4-ethoxycarbonyl)phenylboronic acid
2-Chloro-(4-ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₀BClO₄
MDL Number: MFCD08689531
CAS Number: 913835-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR