4-Hydroxy-3-(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25291G1 AS25291G5
ACTIAS25291G1EA 35 GBP
ACTIAS25291G1 ACTIAS25291G5
4-Hydroxy-3-(trifluoromethyl)benzonitrile
4-Hydroxy-3-(trifluoromethyl)benzonitrile
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
MDL Number: MFCD06797882
CAS Number: 124811-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR