Tubing, rubber, Rotilabo®
Supplier: Carl Roth
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Rotilabo®
ROTH0674.1EA 18.83 GBP
ROTH0674.1 ROTHT983.1 ROTH0677.1 ROTHT984.1 ROTH0676.1 ROTH0675.1 ROTHT981.1 ROTHT982.1
Tubing, rubber, Rotilabo®
Tubing Transfer Tubing
Natural rubber (NR), red.
  • Temperature resistance −10 to +50 °C
  • Not pressure resistant
  • Hardness: 45 Shore A