4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25284G100 AS25284G25 AS25284G5
ACTIAS25284G100EA 0 GBP
ACTIAS25284G100 ACTIAS25284G25 ACTIAS25284G5
4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol
4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
MDL Number: MFCD03093260
CAS Number: 79538-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR