4-(Methylthio)phenyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
024339-1G
024339-5G
FLUO024339-1GEA
0
GBP
FLUO024339-1G
FLUO024339-5G
4-(Methylthio)phenyl isothiocyanate
4-(Methylthio)phenyl isothiocyanate
Formula:
C₈H₇NS₂ Boiling Pt: 127…128 °C (1 mmHg) Melting Pt: 60…62 °C |
MDL Number:
MFCD00041368 CAS Number: 15863-41-9 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...