3-Fluoro-5-methoxybenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4211-5G CA-4211-25G CA-4211-1G
COBBCA-4211-5GEA 0 GBP
COBBCA-4211-5G COBBCA-4211-25G COBBCA-4211-1G
3-Fluoro-5-methoxybenzoic acid
3-Fluoro-5-methoxybenzoic acid
Formula: C₈H₇FO₃
MW: 170.1400 g/mol
MDL Number: MFCD08235092
CAS Number: 176548-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR