3-Ethoxy-2-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4590-1G CA-4590-5G
COBBCA-4590-1GEA 0 GBP
COBBCA-4590-1G COBBCA-4590-5G
3-Ethoxy-2-fluorobenzoic acid
3-Ethoxy-2-fluorobenzoic acid
Formula: C₉H₉FO₃
MW: 184.1600 g/mol
MDL Number: MFCD10699646
CAS Number: 1033201-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR