4-Bromo-2,5-difluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS35740G1 AS35740G5
ACTIAS35740G1EA 0 GBP
ACTIAS35740G1 ACTIAS35740G5
4-Bromo-2,5-difluorobenzoic acid
4-Bromo-2,5-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 237.0000 g/mol
MDL Number: MFCD00844105
CAS Number: 28314-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR