4-Isopropyl-3-thiosemicarbazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018759-2G 018759-5G
FLUO018759-2GEA 0 GBP
FLUO018759-2G FLUO018759-5G
4-Isopropyl-3-thiosemicarbazide
4-Isopropyl-3-thiosemicarbazide
Formula: C₄H₁₁N₃S
MDL Number: MFCD00025144
CAS Number: 13431-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR