Methyl-2,3-dihydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2,3-dihydroxybenzoate

AS20845G100 AS20845G25
ACTIAS20845G100EA 282 GBP
ACTIAS20845G100 ACTIAS20845G25
Methyl-2,3-dihydroxybenzoate
Methyl 2,3-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.1467 g/mol
MDL Number: MFCD00039520
CAS Number: 2411-83-8
EINECS: 219-317-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR