2,6-Difluoro-3-methylbenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2,6-Difluoro-3-methylphenyl)methanol, 2,4-Difluoro-3-(hydroxymethyl)toluene

005755-1G 005755-5G
FLUO005755-1GEA 0 GBP
FLUO005755-1G FLUO005755-5G
2,6-Difluoro-3-methylbenzyl alcohol
2,6-Difluoro-3-methylbenzyl alcohol
Formula: C₈H₈F₂O
MDL Number: MFCD01631335
CAS Number: 261763-40-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR