4-Methoxy-2-methylphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009509-1G 009509-5G
FLUO009509-1GEA 0 GBP
FLUO009509-1G FLUO009509-5G
4-Methoxy-2-methylphenyl isothiocyanate
4-Methoxy-2-methylphenyl isothiocyanate
Formula: C₉H₉NOS
MDL Number: MFCD00060368
CAS Number: 40046-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR