4-Methylbenzyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009521-1G 009521-5G
FLUO009521-1GEA 0 GBP
FLUO009521-1G FLUO009521-5G
4-Methylbenzyl isothiocyanate
4-Methylbenzyl isothiocyanate
Formula: C₉H₉NS
MW: 163.24 g/mol
MDL Number: MFCD00041118
CAS Number: 3694-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR