3-Methylbenzyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009520-1G
009520-5G
FLUO009520-1GEA
0
GBP
FLUO009520-1G
FLUO009520-5G
3-Methylbenzyl isothiocyanate
3-Methylbenzyl isothiocyanate
Formula:
C₉H₉NS |
MDL Number:
MFCD00060410 CAS Number: 3696-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...