2-Methylbenzyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009519-1G
009519-5G
FLUO009519-1GEA
0
GBP
FLUO009519-1G
FLUO009519-5G
2-Methylbenzyl isothiocyanate
2-Methylbenzyl isothiocyanate
Formula:
C₉H₉NS |
MDL Number:
MFCD00060404 CAS Number: 16735-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...