1-Naphthalenemethyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009546-1G
009546-5G
FLUO009546-1GEA
0
GBP
FLUO009546-1G
FLUO009546-5G
1-Naphthalenemethyl isothiocyanate
1-Naphthalenemethyl isothiocyanate
Formula:
C₁₂H₉NS |
MDL Number:
MFCD00060346 CAS Number: 17112-82-2 EINECS: 208-990-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...