2-Methoxyquinoline-3-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5057-5G BB-5057-1G
COBBBB-5057-5GEA 0 GBP
COBBBB-5057-5G COBBBB-5057-1G
2-Methoxyquinoline-3-boronic acid
2-Methoxyquinoline-3-boronic acid
Formula: C₁₀H₁₀BNO₃
MW: 203.0000 g/mol
MDL Number: MFCD09800894
CAS Number: 886853-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR