3-Methyl-4-(methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22344G1 AS22344G5
ACTIAS22344G1EA 0 GBP
ACTIAS22344G1 ACTIAS22344G5
3-Methyl-4-(methylsulphonyl)benzoic acid
3-Methyl-4-(methylsulphonyl)benzoic acid
Formula: C₉H₁₀O₄S
MDL Number: MFCD12828714
CAS Number: 1186663-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR