2-Methyl-4-(methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22339G1 AS22339G25 AS22339G5
ACTIAS22339G1EA 0 GBP
ACTIAS22339G1 ACTIAS22339G25 ACTIAS22339G5
2-Methyl-4-(methylsulphonyl)benzoic acid
2-Methyl-4-(methylsulphonyl)benzoic acid
Formula: C₉H₁₀O₄S
MDL Number: MFCD12828709
CAS Number: 118939-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR