Methyl-3-methyl-2-nitrobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4451-25G CA-4451-5G
COBBCA-4451-25GEA 0 GBP
COBBCA-4451-25G COBBCA-4451-5G
Methyl-3-methyl-2-nitrobenzoate
Methyl 3-methyl-2-nitrobenzoate
Formula: C₉H₉NO₄
MDL Number: MFCD01003947
CAS Number: 5471-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR