Methyl-2-methyl-4-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-methyl-4-nitrobenzoate

AS22455G1 AS22455G25 AS22455G5
ACTIAS22455G1EA 0 GBP
ACTIAS22455G1 ACTIAS22455G25 ACTIAS22455G5
Methyl-2-methyl-4-nitrobenzoate
Methyl 2-methyl-4-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.1700 g/mol
MDL Number: MFCD03618407
CAS Number: 62621-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR