4-Fluoro-3-(methylsulphonyl)benzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22334G1
AS22334G25
AS22334G5
ACTIAS22334G1EA
0
GBP
ACTIAS22334G1
ACTIAS22334G25
ACTIAS22334G5
4-Fluoro-3-(methylsulphonyl)benzoic acid
4-Fluoro-3-(methylsulphonyl)benzoic acid
Formula:
C₈H₇FO₄S |
MDL Number:
MFCD08458082 CAS Number: 158608-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...