3-Fluoro-4-(methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22498G1 AS22498G25 AS22498G5
ACTIAS22498G1EA 0 GBP
ACTIAS22498G1 ACTIAS22498G25 ACTIAS22498G5
3-Fluoro-4-(methylsulphonyl)benzoic acid
3-Fluoro-4-(methylsulphonyl)benzoic acid
Formula: C₈H₇FO₄S
MDL Number: MFCD09743730
CAS Number: 185945-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR