4-(Methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20194G100 AS20194G25
ACTIAS20194G100EA 0 GBP
ACTIAS20194G100 ACTIAS20194G25
4-(Methylsulphonyl)benzoic acid
4-(Methylsulphonyl)benzoic acid
Formula: C₈H₈O₄S
MDL Number: MFCD00007564
CAS Number: 4052-30-6
EINECS: 223-756-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR