3-Chloro-4-(methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22507G1 AS22507G5
ACTIAS22507G1EA 0 GBP
ACTIAS22507G1 ACTIAS22507G5
3-Chloro-4-(methylsulphonyl)benzoic acid
3-Chloro-4-(methylsulphonyl)benzoic acid
Formula: C₈H₇ClO₄S
MDL Number: MFCD13191622
CAS Number: 1197193-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR