4-(3-Fluorophenyl)-3-thiosemicarbazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009240-5G 009240-1G
FLUO009240-5GEA 0 GBP
FLUO009240-5G FLUO009240-1G
4-(3-Fluorophenyl)-3-thiosemicarbazide
4-(3-Fluorophenyl)-3-thiosemicarbazide
Formula: C₇H₈FN₃S
MW: 185.22 g/mol
MDL Number: MFCD00041286
CAS Number: 70619-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR