4-(2,6-Dichlorophenyl)-3-thiosemicarbazide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018540-1G 018540-5G
FLUO018540-1GEA 0 GBP
FLUO018540-1G FLUO018540-5G
4-(2,6-Dichlorophenyl)-3-thiosemicarbazide
4-(2,6-Dichlorophenyl)-3-thiosemicarbazide
Formula: C₇H₇Cl₂N₃S
MDL Number: MFCD00041275
CAS Number: 13207-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR