8-Methylquinoline-5-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 8-Methyl-5-quinolineboronic Acid

BB-5335-5G BB-5335-1G
COBBBB-5335-5GEA 0 GBP
COBBBB-5335-5G COBBBB-5335-1G
8-Methylquinoline-5-boronic acid
8-Methylquinoline-5-boronic acid
Formula: C₁₀H₁₀BNO₂
MW: 187.0000 g/mol
MDL Number: MFCD05256435
CAS Number: 1025010-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR