3-Fluoro-N-methylaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2850-25G
HC-2850-5G
COBBHC-2850-25GEA
0
GBP
COBBHC-2850-25G
COBBHC-2850-5G
3-Fluoro-N-methylaniline hydrochloride
3-Fluoro-N-methylaniline hydrochloride
Formula:
C₇H₉ClFN |
MDL Number:
MFCD12756439 CAS Number: 152814-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...