2-Chloro-N-methylaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2848-5G HC-2848-25G HC-2848-1G
COBBHC-2848-5GEA 0 GBP
COBBHC-2848-5G COBBHC-2848-25G COBBHC-2848-1G
2-Chloro-N-methylaniline hydrochloride
2-Chloro-N-methylaniline hydrochloride
Formula: C₇H₉Cl₂N
MDL Number: MFCD12756437
CAS Number: 1187385-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR