(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024388-1G 24388-1G
FLUO024388-1GEA 0 GBP
FLUO024388-1G FLUO24388-1G
(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride
(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Formula: C₈H₆ClF₃O₂S
MDL Number: MFCD01631926
CAS Number: 127162-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR