2-Fluoro-4-isopropoxyaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1032-5G AN-1032-1G
COBBAN-1032-5GEA 0 GBP
COBBAN-1032-5G COBBAN-1032-1G
2-Fluoro-4-isopropoxyaniline hydrochloride
2-Fluoro-4-isopropoxyaniline hydrochloride
Formula: C₉H₁₃ClFNO
MDL Number: MFCD01632192
CAS Number: 850568-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR