4-Chloro-2-ethoxyaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1076-5G AN-1076-1G
COBBAN-1076-5GEA 0 GBP
COBBAN-1076-5G COBBAN-1076-1G
4-Chloro-2-ethoxyaniline hydrochloride
4-Chloro-2-ethoxyaniline hydrochloride
Formula: C₈H₁₁Cl₂NO
MDL Number: MFCD07783849
CAS Number: 858855-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR