Coconut oil, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

C1758-100G C1758-500G
SIALC1758-100GEA 28 GBP
SIALC1758-100G SIALC1758-500G
Coconut oil, Sigma-Aldrich®
Coconut oil
MDL Number: MFCD00130817
CAS Number: 8001-31-8
EINECS: 232-282-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR