4-(Methoxycarboxy)-1-naphthaleneboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3958-1G BB-3958-250M
COBBBB-3958-1GEA 0 GBP
COBBBB-3958-1G COBBBB-3958-250M
4-(Methoxycarboxy)-1-naphthaleneboronic acid
4-(Methoxycarboxy)-1-naphthaleneboronic acid
Formula: C₁₂H₁₁BO₄
MDL Number: MFCD09475824
CAS Number: 957034-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR