1-Naphthyl boronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Naphthalene-1-boronic acid Pinacol ester, 1-Naphthalenboronic acid, pinacol ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane

021619-5G 021619-25G
FLUO021619-5GEA 0 GBP
FLUO021619-5G FLUO021619-25G
1-Naphthyl boronic acid pinacol ester
1-Naphthyl boronic acid pinacol ester
Formula: C₁₆H₁₉BO₂
MW: 254.1319 g/mol
MDL Number: MFCD05663869
CAS Number: 68716-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR