4-Fluoro-N-(4-methoxybenzyl)benzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4530-5G CA-4530-25G CA-4530-1G
COBBCA-4530-5GEA 0 GBP
COBBCA-4530-5G COBBCA-4530-25G COBBCA-4530-1G
4-Fluoro-N-(4-methoxybenzyl)benzamide
4-Fluoro-N-(4-methoxybenzyl)benzamide
Formula: C₁₅H₁₄FNO₂
MDL Number: MFCD01359589
CAS Number: 346725-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR