3-Benzylaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1119-5G
AN-1119-1G
COBBAN-1119-5GEA
0
GBP
COBBAN-1119-5G
COBBAN-1119-1G
3-Benzylaniline hydrochloride
3-Benzylaniline hydrochloride
Formula:
C₁₃H₁₄ClN |
MDL Number:
MFCD12756412 CAS Number: 6955-30-2 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...