4-Methoxy-N-methylaniline hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2922-5G HC-2922-25G HC-2922-1G
COBBHC-2922-5GEA 0 GBP
COBBHC-2922-5G COBBHC-2922-25G COBBHC-2922-1G
4-Methoxy-N-methylaniline hydrochloride
4-Methoxy-N-methylaniline hydrochloride
Formula: C₈H₁₂ClNO
MW: 173.6400 g/mol
MDL Number: MFCD13196471
CAS Number: 10541-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR