4-Amino-3-fluorophenol hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1025-1G AN-1025-5G
COBBAN-1025-1GEA 0 GBP
COBBAN-1025-1G COBBAN-1025-5G
4-Amino-3-fluorophenol hydrochloride
4-Amino-3-fluorophenol hydrochloride
Formula: C₆H₇ClFNO
MW: 163.5773 g/mol
MDL Number: MFCD01632189
CAS Number: 18266-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR