(3,4-Dichlorophenyl)methanesulphonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13515-1G
013515-1G
FLUO13515-1GEA
0
GBP
FLUO13515-1G
FLUO013515-1G
(3,4-Dichlorophenyl)methanesulphonyl chloride
(3,4-Dichlorophenyl)methanesulphonyl chloride
Formula:
C₇H₅CI₃O₂S MW: 259.54 g/mol |
MDL Number:
MFCD01631929 CAS Number: 85952-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...