(4-Bromophenyl)methanesulphonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Bromobenzylsulphonyl chloride
024384-1G
24384-1G
FLUO024384-1GEA
0
GBP
FLUO024384-1G
FLUO24384-1G
(4-Bromophenyl)methanesulphonyl chloride
(4-Bromophenyl)methanesulphonyl chloride
Formula:
C₇H₆BrClO₂S MW: 269.5433 g/mol |
MDL Number:
MFCD04973454 CAS Number: 53531-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...