(R)-(-)-Glycidyl butyrate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (-)-[(2R)-Oxiran-2-yl]methyl butanoate, (R)-Oxiran-2-ylmethyl butyrate

AS21008G25 AS21008G5
ACTIAS21008G25EA 0 GBP
ACTIAS21008G25 ACTIAS21008G5
(R)-(-)-Glycidyl butyrate
(R)-(-)-Glycidyl butyrate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00075120
CAS Number: 60456-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR