N-Boc-4-bromo-2-fluoroaniline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Boc-4-Bromo-2-fluoro-aniline, tert-Butyl (4-bromo-2-fluorophenyl)carbamate
AS23678G25
AS23678G5
ACTIAS23678G25EA
0
GBP
ACTIAS23678G25
ACTIAS23678G5
N-Boc-4-bromo-2-fluoroaniline
N-Boc-4-bromo-2-fluoroaniline
Formula:
C₁₁H₁₃BrFNO₂ MW: 290.1288 g/mol |
MDL Number:
MFCD07369884 CAS Number: 209958-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...