N-Boc-3-bromo-4-fluoroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23608G1 AS23608G25 AS23608G5
ACTIAS23608G1EA 0 GBP
ACTIAS23608G1 ACTIAS23608G25 ACTIAS23608G5
N-Boc-3-bromo-4-fluoroaniline
N-Boc-3-bromo-4-fluoroaniline
Formula: C₁₁H₁₃BrFNO₂
MW: 290.1300 g/mol
MDL Number: MFCD11975628
CAS Number: 836619-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR