1-Fluoropyridiniumtrifluoromethanesulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Fluoropyridinium triflate, 1-Fluoropyridiniumtriflate, N-Fluoropyridinium trifluoromethanesulphonate

006362-1G 006362-5G
FLUO006362-1GEA 0 GBP
FLUO006362-1G FLUO006362-5G
1-Fluoropyridiniumtrifluoromethanesulphonate
1-Fluoropyridinium trifluoromethanesulphonate
Formula: C₆H₅F₄NO₃S
MDL Number: MFCD00013458
CAS Number: 107263-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR